Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700026977.mm.1 |
NM_145954.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026977.mm.1 |
NM_145954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026977.mm.1 |
TC0700002994.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026979.mm.1 |
NM_145954.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026979.mm.1 |
NM_145954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026979.mm.1 |
TC0700002994.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026982.mm.1 |
NM_145954.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026982.mm.1 |
NM_145954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026982.mm.1 |
TC0700002994.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026980.mm.1 |
NM_145954.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026980.mm.1 |
NM_145954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026980.mm.1 |
TC0700002994.mm.1 |
MTA TC |