Assay Details
| Gene Symbol: Aldh16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145954.1 | NP_666066.1 | 12 - 13 | 1629 | 57 | |
| GenBank mRNA | AK030673.1 | - | 12 - 13 | 1622 | 57 | |
| AK151691.1 | - | 12 - 13 | 1629 | 57 | ||
| AK153129.1 | - | 5 - 6 | 536 | 57 | ||
| AK161198.1 | - | 12 - 13 | 1667 | 57 | ||
| AK164374.1 | - | 12 - 13 | 1675 | 57 | ||
| AK169346.1 | - | 1980 | 57 | |||
| AK170276.1 | - | 12 - 13 | 1668 | 57 | ||
| AK212056.1 | - | 1 - 2 | 72 | 57 | ||
| AK219145.1 | - | 2 - 3 | 182 | 57 | ||
| AK220200.1 | - | 12 - 13 | 1605 | 57 | ||
| BC013548.1 | - | 12 - 13 | 1629 | 4222058 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh16a1
Genomic Map