Assay Details
| Gene Symbol: Nox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172203.2 | NP_757340.1 | 11 - 12 | 1585 | 64 | |
| GenBank mRNA | AF539799.1 | - | 11 - 12 | 1439 | 64 | |
| AK136432.1 | - | 11 - 12 | 1500 | 64 | ||
| AY174116.1 | - | 8 - 9 | 1180 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nox1
Genomic Map