Assay Details
| Gene Symbol: Snx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172277.2 | NP_758481.1 | 2 - 3 | 290 | 70 | |
| GenBank mRNA | AK045061.1 | - | 2 - 3 | 283 | 70 | |
| AK082871.1 | - | 2 - 3 | 283 | 70 | ||
| AK148429.1 | - | 2 - 3 | 283 | 70 | ||
| AK156817.1 | - | 2 - 3 | 236 | 70 | ||
| AK161672.1 | - | 1 - 2 | 224 | 70 | ||
| BC019142.1 | - | 1 - 2 | 212 | 70 | ||
| BC037599.1 | - | 2 - 3 | 316 | 5150754 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snx8
Genomic Map