Assay Details
| Gene Symbol: Ccbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172404.2 | NP_765992.2 | 6 - 7 | 711 | 56 | |
| GenBank mRNA | AK008165.1 | - | 4 - 5 | 488 | 56 | |
| AK031511.1 | - | 6 - 7 | 724 | 56 | ||
| AK042391.1 | - | 5 - 6 | 657 | 56 | ||
| AK088404.1 | - | 5 - 6 | 619 | 56 | ||
| AK163165.1 | - | 5 - 6 | 520 | 56 | ||
| BC016206.1 | - | 6 - 7 | 813 | 4912335 | 56 | |
| BC025124.1 | - | 5 - 6 | 844 | 4486058 | 56 | |
| BC052047.1 | - | 6 - 7 | 711 | 5687475 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccbl1
Genomic Map