Assay Details
| Gene Symbol: Slc38a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172479.3 | NP_766067.2 | 12 - 13 | 1070 | 60 | |
| GenBank mRNA | AK054485.1 | - | 12 - 13 | 1057 | 60 | |
| AK077667.1 | - | 12 - 13 | 1039 | 60 | ||
| AK155928.1 | - | 12 - 13 | 1023 | 60 | ||
| AK158125.1 | - | 11 - 12 | 1191 | 60 | ||
| BC058856.1 | - | 3 - 4 | 328 | 60 | ||
| BC152401.1 | - | 12 - 13 | 1039 | 40057282 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a5
Genomic Map