Assay Details
| Gene Symbol: Mtrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028628.1 | - | 13 - 14 | 1856 | 66 | |
| AK155359.1 | - | 14 - 15 | 2569 | 66 | ||
| AK162978.1 | - | 12 - 13 | 1475 | 66 | ||
| AK163449.1 | - | 13 - 14 | 1838 | 66 | ||
| BC025942.1 | - | 1811 | 5132457 | 66 | ||
| BC031565.1 | - | 13 - 14 | 1852 | 66 | ||
| BC066796.1 | - | 12 - 13 | 1799 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtrr
Genomic Map