Assay Details
Gene Symbol: Srebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011480.3 | NP_035610.1 | 2 - 3 | 576 | 59 | |
GenBank mRNA | AB017337.1 | - | 1 - 2 | 392 | 59 | |
AF374266.1 | - | 1 - 2 | 355 | 59 | ||
AK052628.1 | - | 2 - 3 | 577 | 59 | ||
AK150052.1 | - | 2 - 3 | 565 | 59 | ||
AK154424.1 | - | 2 - 3 | 559 | 59 | ||
AK169607.1 | - | 2 - 3 | 577 | 59 | ||
BC056922.1 | - | 2 - 3 | 558 | 6824948 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
