Assay Details
Gene Symbol: Srebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011480.3 | NP_035610.1 | 6 - 7 | 1224 | 59 | |
GenBank mRNA | AB017337.1 | - | 5 - 6 | 1040 | 59 | |
AF374266.1 | - | 5 - 6 | 988 | 59 | ||
AK052628.1 | - | 6 - 7 | 1225 | 59 | ||
AK154424.1 | - | 6 - 7 | 1192 | 59 | ||
AK169607.1 | - | 6 - 7 | 1225 | 59 | ||
BC006051.1 | - | 3 - 4 | 360 | 59 | ||
BC056922.1 | - | 6 - 7 | 1206 | 6824948 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
