Assay Details
Gene Symbol: Diap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172493.2 | NP_766081.1 | 5 - 6 | 831 | 78 | |
GenBank mRNA | AB713187.1 | - | 6 - 7 | 626 | 78 | |
AK088648.1 | - | 5 - 6 | 765 | 78 | ||
AK153986.1 | - | 5 - 6 | 819 | 78 | ||
AY312280.1 | - | 5 - 6 | 768 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
