Assay Details
| Gene Symbol: Cited4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019563.2 | NP_062509.1 | 1 | 678 | 65 | |
| GenBank mRNA | AF143369.1 | - | 1 | 648 | 65 | |
| AK012136.1 | - | 1 | 670 | 65 | ||
| AK154030.1 | - | 1 | 669 | 65 | ||
| BC025116.1 | - | 2 | 350 | 65 | ||
| BC080723.1 | - | 2 | 435 | 65 | ||
| BC132360.1 | - | 1 | 625 | 40130637 | 65 | |
| BC132362.1 | - | 1 | 625 | 40130639 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252507 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252505 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252502 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252510 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252509 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252506 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252504 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252501 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252508 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252503 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P426768 | AK154030.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P426768 | AF143369.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P426768 | AK012136.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P426768 | BC080723.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P426768 | NM_019563.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P426768 | AK154030.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P426768 | AF143369.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P426768 | AK012136.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P426768 | BC080723.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P426768 | NM_019563.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102539 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P426768 | AK154030.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P426768 | AF143369.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P426768 | AK012136.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P426768 | BC080723.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P426768 | NM_019563.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P426768 | AK154030.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P426768 | AF143369.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P426768 | AK012136.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P426768 | BC080723.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P426768 | NM_019563.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P109099 | AF143369.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23252513 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102539 | - | - |
| Whole Mouse Genome G4122A | A_51_P109099 | AF143369.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P426768 | AK154030.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P426768 | AF143369.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P426768 | AK012136.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P426768 | BC080723.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P426768 | NM_019563.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P426768 | AK154030.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P426768 | AF143369.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P426768 | AK012136.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P426768 | BC080723.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P426768 | NM_019563.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103981 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128500 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102539 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03533722 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04049181 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04093292 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04093293 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04093288 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04093287 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04093297 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04093294 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252507 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252503 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252506 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252504 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252505 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252502 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252510 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252508 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252501 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23252509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23252471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03967553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03723088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03723077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03273875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03912392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04093293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03503297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03939740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03939746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04093294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03533722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03477439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03594869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04093292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03566764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04093295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04093297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03477440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04049181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03273873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03693985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03723092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03939741 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03503304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23252479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03566763 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10507729 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507727 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507728 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425400_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438261_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10507726 | - | - |
| Mouse Genome 430 2.0 Array | 1425400_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1438261_at | - | - |
| Mouse Genome 430 2.0 Array | 1425400_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1438261_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56222_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56222_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400010299.mm.1 | NM_019563.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010299.mm.1 | NM_019563.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010299.mm.1 | TC0400001356.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438261_at | - | - |
| Mouse Expression Set 430 | 1425400_PM_a_at | - | - |
| Mouse Expression Set 430 | 1425400_a_at | - | - |
| Mouse Expression Set 430 | 1438261_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425400_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438261_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438261_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425400_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425400_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438261_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425400_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438261_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 735326 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3130040 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3130040 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003130040 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3130040 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25011.1.118_77-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3130040 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2998934 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2998934 | - | - |

Cited4
Genomic Map