Assay Details
| Gene Symbol: Gtf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025652.3 | NP_079928.2 | 2 - 3 | 422 | 111 | |
| GenBank mRNA | AF391799.1 | - | 2 - 3 | 406 | 111 | |
| AK008247.2 | - | 2 - 3 | 389 | 112 | ||
| AK011845.1 | - | 2 - 3 | 371 | 111 | ||
| AK012797.1 | - | 2 - 3 | 369 | 111 | ||
| AK017234.1 | - | 2 - 3 | 316 | 111 | ||
| AK143627.1 | - | 2 - 3 | 374 | 111 | ||
| BC032292.1 | - | 1 - 2 | 77 | 5374268 | 111 | |
| BC057966.1 | - | 1 - 2 | 634 | 111 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK008247.2,AK012797.1,BC032292.1 transcripts.
Set Membership: |
|

Gtf3a
Genomic Map