Assay Details
Gene Symbol: Brf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025686.2 | NP_079962.1 | 2 - 3 | 271 | 98 | |
GenBank mRNA | AK012680.1 | - | 2 - 3 | 272 | 98 | |
AK017767.1 | - | 2 - 3 | 274 | 98 | ||
AK028228.1 | - | 2 - 3 | 272 | 98 | ||
AK083122.1 | - | 2 - 3 | 272 | 98 | ||
AK151198.1 | - | 2 - 3 | 272 | 98 | ||
AK152842.1 | - | 2 - 3 | 291 | 98 | ||
BC021918.1 | - | 258 | 98 | |||
BC098232.1 | - | 2 - 3 | 252 | 2938123 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25736750 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25736749 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25736751 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25736748 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P507974 | BC098232.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P507974 | AK151198.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P507974 | AK152842.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P507974 | BC021918.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P507974 | AK012680.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P507974 | BC098232.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P507974 | AK151198.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P507974 | AK152842.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P507974 | BC021918.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P507974 | AK012680.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392528 | AJ409501.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392528 | AK135780.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392528 | AK145507.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392528 | AK158003.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392528 | AK162243.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392528 | AK171941.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392528 | AY036118.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392528 | AY248756.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392528 | BC042411.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392528 | BC115496.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392528 | BC115497.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 434124 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 152314 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 134863 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106208 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166882 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P507974 | BC098232.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P507974 | AK151198.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P507974 | AK152842.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P507974 | BC021918.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P507974 | AK012680.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P507974 | BC098232.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P507974 | AK151198.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P507974 | AK152842.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P507974 | BC021918.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P507974 | AK012680.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P507974 | BC098232.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P507974 | AK151198.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P507974 | AK152842.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P507974 | BC021918.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P507974 | AK012680.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P507974 | BC098232.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P507974 | AK151198.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P507974 | AK152842.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P507974 | BC021918.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P507974 | AK012680.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736770 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736743 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736739 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736767 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736763 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736755 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736742 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736747 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736768 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25736751 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166882 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106208 | - | - |
Whole Mouse Genome G4122A | A_51_P507974 | BC098232.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P507974 | AK151198.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P507974 | AK152842.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P507974 | BC021918.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P507974 | AK012680.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P507974 | BC098232.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P507974 | AK151198.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P507974 | AK152842.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P507974 | BC021918.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P507974 | AK012680.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130163 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106208 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166882 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07318057 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07318060 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07312243 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07318062 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25736751 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25736748 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25736750 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25736749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25736770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07021791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07165034 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736782 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736726 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07319579 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07318062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736743 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736745 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07060558 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07318064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736778 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736754 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736751 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736725 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736739 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06786948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736734 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07021786 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736724 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736764 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736773 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06900917 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736760 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736742 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736748 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736736 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736756 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736767 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736744 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25736757 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10577953 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10577950 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10577951 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10577952 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10577949 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429561_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10577948 | - | - |
Mouse Genome 430 2.0 Array | 1429561_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1429561_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66653_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66653_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800015508.mm.1 | NM_025686.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800015508.mm.1 | NM_025686.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800015508.mm.1 | TC0800001951.mm.1 | MTA TC |
Mouse Expression Set 430 | 1429561_PM_at | - | - |
Mouse Expression Set 430 | 1429561_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429561_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429561_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 499249 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6900167 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006900167 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1258583 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1258583 | - | - |
mouse-6 v1.1 (Illumina) | scl066653.1_40-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6900167 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl066653.1_40-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900167 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1258583 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1258583 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066653.1_40-S | - | - |
