Assay Details
| Gene Symbol: Tcf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037877.3 | NP_001032966.1 | 14 - 15 | 1794 | 101 | |
| NM_001037878.3 | NP_001032967.1 | 14 - 15 | 1794 | 101 | ||
| NM_001286362.1 | NP_001273291.1 | 13 - 14 | 1719 | 101 | ||
| NM_001286363.1 | NP_001273292.1 | 13 - 14 | 1719 | 101 | ||
| NM_025804.3 | NP_080080.2 | 14 - 15 | 1794 | 101 | ||
| GenBank mRNA | AK003180.1 | - | 14 - 15 | 1681 | 101 | |
| AK013754.1 | - | 14 - 15 | 1483 | 101 | ||
| AK045397.1 | - | 13 - 14 | 1647 | 101 | ||
| AK050274.1 | - | 14 - 15 | 1720 | 101 | ||
| AK075728.1 | - | 14 - 15 | 1644 | 101 | ||
| AK076151.1 | - | 8 - 9 | 881 | 101 | ||
| AK088360.1 | - | 14 - 15 | 1682 | 101 | ||
| AK088602.1 | - | 1 - 2 | 333 | 101 | ||
| AK089964.1 | - | 14 - 15 | 1683 | 101 | ||
| AK122430.1 | - | 14 - 15 | 1649 | 101 | ||
| AK148082.1 | - | 14 - 15 | 1656 | 101 | ||
| AK150918.1 | - | 14 - 15 | 1675 | 101 | ||
| AK150942.1 | - | 14 - 15 | 1668 | 101 | ||
| AK151143.1 | - | 14 - 15 | 1668 | 101 | ||
| AK151558.1 | - | 14 - 15 | 1674 | 101 | ||
| AK152178.1 | - | 14 - 15 | 1668 | 101 | ||
| AK152494.1 | - | 14 - 15 | 1668 | 101 | ||
| AK161627.1 | - | 14 - 15 | 1670 | 101 | ||
| AK167837.1 | - | 14 - 15 | 1668 | 101 | ||
| AK168167.1 | - | 14 - 15 | 1668 | 101 | ||
| BC016130.1 | - | 14 - 15 | 1656 | 101 | ||
| BC025071.1 | - | 14 - 15 | 1655 | 3498003 | 101 | |
| BC046768.1 | - | 14 - 15 | 1752 | 5707312 | 101 | |
| EU624323.1 | - | 13 - 14 | 1557 | 101 | ||
| U94988.1 | - | 14 - 15 | 1655 | 101 | ||
More Information
Set Membership: |
|

Tcf25
Genomic Map