Assay Details
| Gene Symbol: Ift80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026641.2 | NP_080917.1 | 12 - 13 | 1493 | 76 | |
| GenBank mRNA | AK042231.1 | - | 12 - 13 | 1418 | 76 | |
| AK147305.1 | - | 11 - 12 | 5020 | 76 | ||
| BC013814.1 | - | 7 - 8 | 782 | 76 | ||
| BC034101.1 | - | 12 - 13 | 1388 | 4501451 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ift80
Genomic Map