Assay Details
Gene Symbol: Mllt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286158.1 | NP_001273087.1 | 3 - 4 | 316 | 88 | |
NM_027326.3 | NP_081602.3 | 7 - 8 | 1643 | 88 | ||
NM_029931.3 | NP_084207.2 | 3 - 4 | 316 | 88 | ||
GenBank mRNA | AF333960.1 | - | 6 - 7 | 1304 | 88 | |
AK008707.1 | - | 3 - 4 | 278 | 88 | ||
AK019458.1 | - | 3 - 4 | 308 | 88 | ||
AK147009.1 | - | 3 - 4 | 230 | 88 | ||
BC021420.1 | - | 8 - 9 | 1482 | 5012671 | 88 | |
BU614509.1 | - | 3 - 4 | 444 | 88 | ||
BY713607.1 | - | 3 - 4 | 304 | 88 | ||
DV051568.1 | - | 326 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
