Assay Details
| Gene Symbol: Dmrtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027732.2 | NP_082008.1 | 5 - 6 | 755 | 64 | |
| GenBank mRNA | AF542046.1 | - | 5 - 6 | 752 | 64 | |
| AK017020.1 | - | 5 - 6 | 753 | 64 | ||
| BC100434.1 | - | 5 - 6 | 755 | 30918218 | 64 | |
| BC138203.1 | - | 5 - 6 | 686 | 8861222 | 64 | |
| BC145988.1 | - | 5 - 6 | 686 | 40131291 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dmrtc2
Genomic Map