Assay Details
| Gene Symbol: Ppp6r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026813.1 | NP_081089.1 | 4 - 5 | 587 | 115 | |
| NM_027805.2 | NP_082081.1 | 4 - 5 | 587 | 115 | ||
| GenBank mRNA | AK018403.1 | - | 4 - 5 | 507 | 115 | |
| AK083335.1 | - | 4 - 5 | 579 | 115 | ||
| AK084684.1 | - | 4 - 5 | 588 | 115 | ||
| BC024879.1 | - | 3 - 4 | 390 | 4011449 | 115 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK083335.1,AK084684.1,BC024879.1,NM_026813.1,NM_027805.2 transcripts.
Set Membership: |
|

Ppp6r2
Genomic Map