Assay Details
| Gene Symbol: Shroom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290789.1 | NP_001277718.1 | 4 - 5 | 1712 | 100 | |
| NM_027917.3 | NP_082193.2 | 5 - 6 | 1901 | 100 | ||
| GenBank mRNA | AK004934.1 | - | 5 - 6 | 1828 | 100 | |
| AK134332.1 | - | 5 - 6 | 1829 | 100 | ||
| BC005761.1 | - | 4 - 5 | 1687 | 3601742 | 100 | |
| BC031598.1 | - | 4 - 5 | 1604 | 3497170 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Shroom1
Genomic Map