Assay Details
Gene Symbol: Ddx54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028041.2 | NP_082317.1 | 3 - 4 | 379 | 106 | |
GenBank mRNA | AF319547.1 | - | 3 - 4 | 739 | 106 | |
AK028274.1 | - | 3 - 4 | 393 | 106 | ||
AK031534.1 | - | 3 - 4 | 389 | 106 | ||
AK031679.1 | - | 3 - 4 | 397 | 106 | ||
AK147715.1 | - | 3 - 4 | 394 | 106 | ||
AK155138.1 | - | 3 - 4 | 391 | 106 | ||
AK167603.1 | - | 3 - 4 | 393 | 106 | ||
AK168455.1 | - | 3 - 4 | 393 | 106 | ||
AK169255.1 | - | 3 - 4 | 391 | 106 | ||
AK170967.1 | - | 3 - 4 | 391 | 106 | ||
BC043699.1 | - | 3 - 4 | 379 | 5344158 | 106 | |
BC062920.1 | - | 3 - 4 | 382 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
