Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700018940.mm.1 |
NM_001142744.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018940.mm.1 |
NM_001142744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018940.mm.1 |
NM_001142745.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018940.mm.1 |
NM_001142745.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018940.mm.1 |
NM_028476.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018940.mm.1 |
NM_028476.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018940.mm.1 |
TC1700001986.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700018938.mm.1 |
NM_001142744.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018938.mm.1 |
NM_001142744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018938.mm.1 |
NM_001142745.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018938.mm.1 |
NM_001142745.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018938.mm.1 |
NM_028476.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018938.mm.1 |
NM_028476.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018938.mm.1 |
TC1700001986.mm.1 |
MTA TC |