| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800023324.mm.1 |
NM_028883.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023324.mm.1 |
NM_028883.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023324.mm.1 |
TC0800003030.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023321.mm.1 |
NM_028883.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023321.mm.1 |
NM_028883.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023321.mm.1 |
TC0800003030.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023322.mm.1 |
NM_028883.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023322.mm.1 |
NM_028883.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023322.mm.1 |
TC0800003030.mm.1 |
MTA TC |