Assay Details
| Gene Symbol: Pck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028994.2 | NP_083270.1 | 4 - 5 | 886 | 77 | |
| GenBank mRNA | AK007429.1 | - | 1 - 2 | 80 | 77 | |
| AK030501.1 | - | 4 - 5 | 887 | 77 | ||
| AK031612.1 | - | 4 - 5 | 731 | 77 | ||
| AK034927.1 | - | 4 - 5 | 765 | 77 | ||
| AK147972.1 | - | 4 - 5 | 740 | 77 | ||
| AK165094.1 | - | 4 - 5 | 764 | 77 | ||
| AK170242.1 | - | 4 - 5 | 740 | 77 | ||
| BC010318.1 | - | 4 - 5 | 749 | 3596227 | 77 | |
| BC011126.1 | - | 4 - 5 | 746 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pck2
Genomic Map