Assay Details
| Gene Symbol: Pck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028994.2 | NP_083270.1 | 6 - 7 | 1235 | 71 | |
| GenBank mRNA | AK007429.1 | - | 3 - 4 | 429 | 71 | |
| AK030501.1 | - | 6 - 7 | 1236 | 71 | ||
| AK031612.1 | - | 6 - 7 | 1080 | 71 | ||
| AK034927.1 | - | 6 - 7 | 1114 | 71 | ||
| AK147972.1 | - | 6 - 7 | 1089 | 71 | ||
| AK165094.1 | - | 6 - 7 | 1113 | 71 | ||
| AK170242.1 | - | 6 - 7 | 1089 | 71 | ||
| AK189946.1 | - | 1 - 2 | 61 | 71 | ||
| BC010318.1 | - | 6 - 7 | 1098 | 3596227 | 71 | |
| BC011126.1 | - | 5 - 6 | 1397 | 71 | ||
| BC028326.1 | - | 1 - 2 | 36 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pck2
Genomic Map