Assay Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029186.2 | NP_083462.2 | 4 - 5 | 884 | 82 | |
| GenBank mRNA | AK015988.1 | - | 1 - 2 | 241 | 82 | |
| AK031554.1 | - | 4 - 5 | 830 | 82 | ||
| AK151797.1 | - | 4 - 5 | 859 | 82 | ||
| AK161305.1 | - | 4 - 5 | 840 | 82 | ||
| AK170247.1 | - | 831 | 82 | |||
| BC026385.1 | - | 2 - 3 | 1012 | 82 | ||
| BC031169.1 | - | 4 - 5 | 829 | 82 | ||
| BC049142.1 | - | 1 - 2 | 170 | 82 | ||
| BC058746.1 | - | 4 - 5 | 830 | 6399588 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem180
Genomic Map