Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700023462.mm.1 |
NM_001171004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023462.mm.1 |
NM_001171004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023462.mm.1 |
NM_001171005.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023462.mm.1 |
NM_001171005.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023462.mm.1 |
NM_029239.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023462.mm.1 |
NM_029239.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023462.mm.1 |
TC1700002562.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700023461.mm.1 |
NM_001171004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023461.mm.1 |
NM_001171004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023461.mm.1 |
NM_001171005.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023461.mm.1 |
NM_001171005.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023461.mm.1 |
NM_029239.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023461.mm.1 |
NM_029239.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023461.mm.1 |
TC1700002562.mm.1 |
MTA TC |