Assay Details
| Gene Symbol: Chmp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029362.3 | NP_083638.1 | 3 - 4 | 637 | 60 | |
| GenBank mRNA | AK156473.1 | - | 3 - 4 | 597 | 60 | |
| AK159193.1 | - | 3 - 4 | 599 | 60 | ||
| AK164550.1 | - | 526 | 60 | |||
| AK196942.1 | - | 2 - 3 | 146 | 60 | ||
| BC006905.1 | - | 3 - 4 | 353 | 60 | ||
| BC011429.1 | - | 3 - 4 | 614 | 3485793 | 60 | |
| BC059279.1 | - | 3 - 4 | 589 | 4019739 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chmp4b
Genomic Map