Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300017651.mm.1 |
NM_029447.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017651.mm.1 |
NM_029447.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017651.mm.1 |
TC1300002594.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300017650.mm.1 |
NM_029447.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017650.mm.1 |
NM_029447.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017650.mm.1 |
TC1300002594.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300017649.mm.1 |
NM_029447.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017649.mm.1 |
NM_029447.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017649.mm.1 |
TC1300002594.mm.1 |
MTA TC |