Assay Details
| Gene Symbol: Ctsq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029636.3 | NP_083912.2 | 1 - 2 | 31 | 83 | |
| GenBank mRNA | AF456461.1 | - | 1 - 2 | 62 | 83 | |
| AY014776.1 | - | 1 - 2 | 31 | 83 | ||
| BC099415.1 | - | 1 - 2 | 38 | 30786803 | 83 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF456461.1,AY014776.1,BC099415.1,NM_029636.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88829 | AY014776.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88829 | BC099415.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88829 | NM_029636.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88829 | AK005416.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88829 | AY014776.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88829 | BC099415.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88829 | NM_029636.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P88829 | AK005416.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210878 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 255805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49664 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148030 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88829 | AY014776.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88829 | BC099415.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88829 | NM_029636.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88829 | AK005416.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88829 | AY014776.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88829 | BC099415.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88829 | NM_029636.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P88829 | AK005416.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088392 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148030 | - | - |
| Whole Mouse Genome G4122A | A_52_P88829 | AY014776.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88829 | BC099415.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88829 | NM_029636.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88829 | AK005416.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88829 | AY014776.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88829 | BC099415.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88829 | NM_029636.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P88829 | AK005416.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136943 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148030 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10491495 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10491475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10745054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10491496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10756104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10707086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10707085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088346 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088336 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10409902 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409903 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409901 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409900 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409898 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409897 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409899 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409904 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422289_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10409896 | - | - |
| Mouse Genome 430 2.0 Array | 1422289_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422289_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 104002_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 104002_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300013789.mm.1 | NM_029636.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013789.mm.1 | NM_029636.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013789.mm.1 | TC1300002146.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300013790.mm.1 | NM_029636.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013790.mm.1 | NM_029636.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013790.mm.1 | TC1300002146.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422289_PM_a_at | - | - |
| Mouse Expression Set 430 | 1422289_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422289_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422289_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422289_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422289_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 501462 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3190592 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003190592 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2776341 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2776341 | - | - |
| mouse-6 v1.1 (Illumina) | scl43860.8.1_146-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3190592 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl43860.8.1_146-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190592 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2981137 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2776341 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2981137 | - | - |

Ctsq
Genomic Map