Assay Details
| Gene Symbol: 6430548M08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163760.1 | NP_001157232.1 | 1 - 2 | 146 | 97 | |
| GenBank mRNA | BC027126.1 | - | 1 - 2 | 146 | 5359842 | 97 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC027126.1,NM_001163760.1 transcripts.
Set Membership: |
|

6430548M08Rik
Genomic Map