Assay Details
| Gene Symbol: Inpp5j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172439.3 | NP_766027.2 | 6 - 7 | 1842 | 85 | |
| GenBank mRNA | AK034272.1 | - | 6 - 7 | 1843 | 85 | |
| AK136908.1 | - | 6 - 7 | 1829 | 85 | ||
| AK165718.1 | - | 5 - 6 | 2370 | 85 | ||
| AK170114.1 | - | 6 - 7 | 1808 | 85 | ||
| AK171029.1 | - | 6 - 7 | 1803 | 85 | ||
| BC022915.1 | - | 6 - 7 | 1795 | 85 | ||
| BC046561.1 | - | 5 - 6 | 655 | 85 | ||
| BC051452.1 | - | 6 - 7 | 1811 | 85 | ||
| BC131634.1 | - | 6 - 7 | 1837 | 40096940 | 85 | |
| BC131635.1 | - | 6 - 7 | 1834 | 40096943 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inpp5j
Genomic Map