Assay Details
Gene Symbol: Nek11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172461.3 | NP_766049.2 | 4 - 5 | 693 | 120 | |
GenBank mRNA | AK030042.1 | - | 4 - 5 | 652 | 120 | |
AK043543.1 | - | 4 - 5 | 932 | 120 | ||
AK054237.1 | - | 4 - 5 | 587 | 120 | ||
BC119588.1 | - | 3 - 4 | 527 | 40058889 | 120 | |
BC119589.1 | - | 3 - 4 | 527 | 40058890 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
