Assay Details
| Gene Symbol: Dis3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001295.2 | NP_001001295.2 | 5 - 6 | 864 | 65 | |
| NM_001177784.1 | NP_001171255.1 | 5 - 6 | 825 | 65 | ||
| NM_172519.3 | NP_766107.1 | 5 - 6 | 768 | 65 | ||
| GenBank mRNA | AK029974.1 | - | 5 - 6 | 769 | 65 | |
| AK032451.1 | - | 5 - 6 | 773 | 65 | ||
| AK035357.1 | - | 5 - 6 | 764 | 65 | ||
| AK077031.1 | - | 5 - 6 | 828 | 65 | ||
| AK150851.1 | - | 5 - 6 | 814 | 65 | ||
| AK151780.1 | - | 5 - 6 | 814 | 65 | ||
| AK153433.1 | - | 5 - 6 | 814 | 65 | ||
| AK173310.1 | - | 5 - 6 | 798 | 65 | ||
| BC056939.1 | - | 5 - 6 | 797 | 5708685 | 65 | |
More Information
Set Membership: |
|

Dis3l
Genomic Map