Assay Details
| Gene Symbol: She | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172530.3 | NP_766118.3 | 2 - 3 | 868 | 79 | |
| GenBank mRNA | AK034662.1 | - | 2 - 3 | 879 | 79 | |
| AK157580.1 | - | 2 - 3 | 868 | 79 | ||
| BC109362.1 | - | 3 - 4 | 384 | 40058813 | 79 | |
| BC109363.1 | - | 2 - 3 | 840 | 40058814 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

She
Genomic Map