Assay Details
| Gene Symbol: Sema6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052232.1 | - | 1 - 2 | 169 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK052232.1 transcripts.
Set Membership: |
|

Sema6d
Genomic Map