| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00018622.mm.1 |
NM_001167995.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018622.mm.1 |
NM_001167995.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018622.mm.1 |
TC0X00002990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00018625.mm.1 |
NM_001167994.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018625.mm.1 |
NM_001167994.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018625.mm.1 |
NM_001167995.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018625.mm.1 |
NM_001167995.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018625.mm.1 |
NM_172540.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018625.mm.1 |
NM_172540.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018625.mm.1 |
TC0X00002990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00018626.mm.1 |
NM_001167994.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018626.mm.1 |
NM_001167994.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018626.mm.1 |
NM_001167995.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018626.mm.1 |
NM_001167995.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018626.mm.1 |
NM_172540.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018626.mm.1 |
NM_172540.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018626.mm.1 |
TC0X00002990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00018631.mm.1 |
NM_001167994.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018631.mm.1 |
NM_001167994.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018631.mm.1 |
NM_001167995.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018631.mm.1 |
NM_001167995.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018631.mm.1 |
NM_172540.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018631.mm.1 |
NM_172540.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018631.mm.1 |
TC0X00002990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00018627.mm.1 |
NM_001167994.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018627.mm.1 |
NM_001167994.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018627.mm.1 |
TC0X00002990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00018628.mm.1 |
NM_001167994.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018628.mm.1 |
NM_001167994.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018628.mm.1 |
NM_001167995.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018628.mm.1 |
NM_001167995.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018628.mm.1 |
NM_172540.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018628.mm.1 |
NM_172540.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00018628.mm.1 |
TC0X00002990.mm.1 |
MTA TC |