Assay Details
| Gene Symbol: Polrmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172551.3 | NP_766139.2 | 18 - 19 | 3434 | 54 | |
| GenBank mRNA | AK003792.2 | - | 9 - 10 | 1172 | 54 | |
| AK030039.1 | - | 17 - 18 | 3216 | 54 | ||
| AK042071.1 | - | 16 - 17 | 2991 | 54 | ||
| AK047545.1 | - | 14 - 15 | 3959 | 54 | ||
| AK053356.1 | - | 18 - 19 | 3425 | 54 | ||
| AK085666.1 | - | 18 - 19 | 3427 | 54 | ||
| AK128956.1 | - | 16 - 17 | 3311 | 54 | ||
| AK154181.1 | - | 17 - 18 | 3471 | 54 | ||
| AK154729.1 | - | 18 - 19 | 3416 | 54 | ||
| BC110697.1 | - | 18 - 19 | 3410 | 30356222 | 54 | |
| BC117939.1 | - | 16 - 17 | 3611 | 54 | ||
| BC117940.1 | - | 18 - 19 | 3427 | 40097816 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polrmt
Genomic Map