Assay Details
Gene Symbol: Rhbdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167680.1 | NP_001161152.1 | 3 - 4 | 453 | 72 | |
NM_172572.3 | NP_766160.2 | 3 - 4 | 667 | 72 | ||
GenBank mRNA | AK028872.1 | - | 3 - 4 | 668 | 72 | |
AK042365.1 | - | 3 - 4 | 449 | 72 | ||
AK154639.1 | - | 3 - 4 | 422 | 72 | ||
AK171085.1 | - | 3 - 4 | 449 | 72 | ||
AY167043.1 | - | 3 - 4 | 453 | 72 | ||
BC052182.1 | - | 3 - 4 | 401 | 6335022 | 72 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK042365.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
