Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100039058.mm.1 |
NM_001167680.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039058.mm.1 |
NM_001167680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039058.mm.1 |
NM_172572.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039058.mm.1 |
NM_172572.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039058.mm.1 |
TC1100004122.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100039057.mm.1 |
NM_001167680.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039057.mm.1 |
NM_001167680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039057.mm.1 |
NM_172572.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039057.mm.1 |
NM_172572.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039057.mm.1 |
TC1100004122.mm.1 |
MTA TC |