Assay Details
| Gene Symbol: Fcho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172591.3 | NP_766179.2 | 6 - 7 | 940 | 108 | |
| GenBank mRNA | AK031041.1 | - | 6 - 7 | 943 | 108 | |
| AK142282.1 | - | 6 - 7 | 626 | 108 | ||
| AK146566.1 | - | 6 - 7 | 674 | 108 | ||
| AK164135.1 | - | 6 - 7 | 686 | 108 | ||
| BC019933.1 | - | 7 - 8 | 676 | 108 | ||
| BC047122.1 | - | 7 - 8 | 676 | 108 | ||
| BC052456.1 | - | 6 - 7 | 637 | 6830607 | 108 | |
| BC053718.1 | - | 6 - 7 | 656 | 108 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK164135.1 transcripts.
Set Membership: |
|

Fcho2
Genomic Map