Assay Details
Gene Symbol: Tdrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253755.1 | NP_001240684.1 | 9 - 10 | 1237 | 82 | |
NM_172605.3 | NP_766193.3 | 9 - 10 | 1237 | 82 | ||
GenBank mRNA | AK032892.1 | - | 9 - 10 | 1182 | 82 | |
AK033390.1 | - | 9 - 10 | 1182 | 82 | ||
BC005670.1 | - | 9 - 10 | 1138 | 3153274 | 82 | |
BC066144.1 | - | 9 - 10 | 1185 | 30435963 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
