| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700004045.mm.1 |
NM_172617.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004045.mm.1 |
NM_172617.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004045.mm.1 |
TC1700000472.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004050.mm.1 |
NM_172617.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004050.mm.1 |
NM_172617.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004050.mm.1 |
TC1700000472.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004046.mm.1 |
NM_172617.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004046.mm.1 |
NM_172617.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004046.mm.1 |
TC1700000472.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004048.mm.1 |
NM_172617.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004048.mm.1 |
NM_172617.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004048.mm.1 |
TC1700000472.mm.1 |
MTA TC |