Assay Details
| Gene Symbol: Clic5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172621.2 | NP_766209.1 | 3 - 4 | 628 | 58 | |
| GenBank mRNA | AK040195.1 | - | 2 - 3 | 266 | 58 | |
| AK046522.1 | - | 3 - 4 | 617 | 58 | ||
| AK156849.1 | - | 3 - 4 | 469 | 58 | ||
| BC064037.1 | - | 3 - 4 | 613 | 1227040 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clic5
Genomic Map