Assay Details
Gene Symbol: Mppe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172630.2 | NP_766218.1 | 4 - 5 | 1025 | 98 | |
GenBank mRNA | AK041262.1 | - | 4 - 5 | 1026 | 98 | |
AK154265.1 | - | 4 - 5 | 879 | 98 | ||
AK158446.1 | - | 4 - 5 | 748 | 98 | ||
AK172000.1 | - | 4 - 5 | 864 | 98 | ||
AK172496.1 | - | 4 - 5 | 780 | 98 | ||
BC045146.1 | - | 4 - 5 | 651 | 6306245 | 98 | |
BC086456.1 | - | 4 - 5 | 698 | 4949311 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790837 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790832 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790834 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790836 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790831 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790833 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790838 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139694 | AK041262.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139694 | AK154265.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139694 | NM_172630.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139694 | AK172000.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564915 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190700 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163915 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510838 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 387357 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108201 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404845 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200000 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 619870 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875047 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 59614 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 140579 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125872 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127721 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139694 | AK041262.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139694 | AK154265.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139694 | NM_172630.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139694 | AK172000.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790826 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790844 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790855 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790824 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790820 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790865 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790842 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790831 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790819 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790850 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790843 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790822 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790860 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790833 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790828 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790830 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790862 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790836 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790852 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790858 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790840 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790846 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790839 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790864 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790837 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31790848 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09036024 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125872 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127721 | - | - |
Whole Mouse Genome G4122A | A_51_P136361 | BC086456.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P136361 | BC045146.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P136361 | AK041262.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P136361 | NM_172630.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P136361 | NM_010307.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P344741 | BC086456.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P344741 | NM_010307.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P344741 | NM_177137.4 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139694 | AK041262.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139694 | AK154265.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139694 | NM_172630.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139694 | AK172000.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134722 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125109 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127721 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125872 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13807891 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13840588 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13840596 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13840593 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13880665 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13840603 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13840590 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790834 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790838 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790837 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790836 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790831 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790833 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31790832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31790855 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790817 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13871278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13760700 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13812873 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790819 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790846 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790855 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790826 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790842 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13645751 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790858 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790833 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790789 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790865 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790807 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790868 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790871 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13839835 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790864 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790847 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790836 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790852 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790804 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790866 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13880665 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790809 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13840601 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790810 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790822 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790799 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790824 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790840 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13645744 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790853 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790845 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790837 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13895533 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790839 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790869 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790820 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13895536 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790790 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13850829 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31790828 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13910847 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13851977 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10459525 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10459523 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10459526 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10459527 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10459522 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10459524 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10459519 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10459521 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10459520 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10459529 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10459528 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1440880_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10459518 | - | - |
Mouse Genome 430 2.0 Array | 1440880_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1440880_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1800010986.mm.1 | NM_172630.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800010986.mm.1 | NM_172630.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800010986.mm.1 | TC1800001500.mm.1 | MTA TC |
Mouse Expression Set 430 | 1440880_PM_at | - | - |
Mouse Expression Set 430 | 1440880_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440880_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440880_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 921152 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5080364 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 7050739 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5080364 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005080364 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0007050739 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2733214 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2678912 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2733214 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2678912 | - | - |
mouse-6 v1.1 (Illumina) | scl51323.13.1_30-S | - | - |
mouse-6 v1.1 (Illumina) | scl0002237.1_23-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 7050739 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5080364 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl51323.13.1_30-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002237.1_23-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7050739 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5080364 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2812244 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2733214 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2812244 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2733214 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl51323.13.1_30-S | - | - |
