Assay Details
| Gene Symbol: Mppe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172630.2 | NP_766218.1 | 6 - 7 | 1198 | 68 | |
| GenBank mRNA | AK041262.1 | - | 6 - 7 | 1199 | 68 | |
| AK154265.1 | - | 6 - 7 | 1052 | 68 | ||
| AK172496.1 | - | 6 - 7 | 953 | 68 | ||
| BC045146.1 | - | 6 - 7 | 824 | 6306245 | 68 | |
| BC086456.1 | - | 6 - 7 | 871 | 4949311 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mppe1
Genomic Map