Assay Details
Gene Symbol: Kctd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172650.2 | NP_766238.1 | 6 - 7 | 627 | 83 | |
GenBank mRNA | AK033491.1 | - | 6 - 7 | 628 | 83 | |
AK034380.1 | - | 6 - 7 | 627 | 83 | ||
AK049362.1 | - | 6 - 7 | 625 | 83 | ||
AK087407.1 | - | 1 - 2 | 739 | 83 | ||
AK087865.1 | - | 6 - 7 | 628 | 83 | ||
AK139700.1 | - | 6 - 7 | 627 | 83 | ||
AK215667.1 | - | 1 - 2 | 40 | 83 | ||
BC061458.1 | - | 6 - 7 | 615 | 83 | ||
BC079581.1 | - | 6 - 7 | 614 | 5702630 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
