Assay Details
| Gene Symbol: Tmem194b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142647.1 | NP_001136119.1 | 8 - 9 | 1186 | 72 | |
| NR_024590.1 | 8 - 9 | 1140 | 72 | |||
| GenBank mRNA | AK030361.1 | - | 7 - 8 | 1041 | 72 | |
| AK036693.1 | - | 8 - 9 | 1100 | 72 | ||
| AK048484.1 | - | 8 - 9 | 1097 | 72 | ||
| AK086801.1 | - | 4 - 5 | 583 | 72 | ||
| AK136177.1 | - | 4 - 5 | 674 | 72 | ||
| AK170577.1 | - | 3 - 4 | 2490 | 72 | ||
| BC028796.1 | - | 5 - 6 | 612 | 72 | ||
| BC094370.1 | - | 3 - 4 | 2130 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489736 | BC094370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489736 | BC016122.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489736 | AK030361.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489736 | NR_024590.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489736 | NM_001142647.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489736 | BC094370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489736 | BC016122.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489736 | AK030361.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489736 | NR_024590.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489736 | NM_001142647.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P51854 | BC028796.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P51854 | BC094370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P51854 | AK136177.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P51854 | AK170577.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P51854 | AK086801.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P51854 | BC028796.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P51854 | BC094370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P51854 | AK136177.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P51854 | AK170577.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P51854 | AK086801.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513581 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691702 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 305959 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348797 | AK014276.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348797 | AK016966.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348797 | AK018101.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348797 | AK032498.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348797 | BC018481.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348797 | NM_025755.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616319 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343109 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 599810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64040 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 769615 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767523 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 114261 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 171503 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145547 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489736 | BC094370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489736 | BC016122.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489736 | AK030361.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489736 | NR_024590.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489736 | NM_001142647.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489736 | BC094370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489736 | BC016122.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489736 | AK030361.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489736 | NR_024590.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489736 | NM_001142647.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P51854 | BC028796.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P51854 | BC094370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P51854 | AK136177.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P51854 | AK170577.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P51854 | AK086801.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P51854 | BC028796.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P51854 | BC094370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P51854 | AK136177.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P51854 | AK170577.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P51854 | AK086801.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P51854 | BC028796.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P51854 | BC094370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P51854 | AK136177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P51854 | AK170577.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P51854 | AK086801.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P51854 | BC028796.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P51854 | BC094370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P51854 | AK136177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P51854 | AK170577.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P51854 | AK086801.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489736 | BC094370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489736 | BC016122.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489736 | AK030361.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489736 | NR_024590.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489736 | NM_001142647.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489736 | BC094370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489736 | BC016122.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489736 | AK030361.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489736 | NR_024590.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489736 | NM_001142647.1 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00959389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00383587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00383586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00947429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00383584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00966579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00326301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00383593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00383595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00555203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00032029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01098316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00395231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00959390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00053283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00322945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01136297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20228122 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10346228 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346229 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346234 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346230 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346232 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346233 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346227 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346226 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346231 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346225 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455553_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10346224 | - | - |
| Mouse Genome 430 2.0 Array | 1455553_at | - | - |
| Mouse Genome 430 2.0 Array | 1445530_at | - | - |
| Mouse Genome 430 2.0 Array | 1455553_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1445530_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100002951.mm.1 | NM_001142647.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100002951.mm.1 | NM_001142647.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100002951.mm.1 | NR_024590.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100002951.mm.1 | NR_024590.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100002951.mm.1 | TC0100000380.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100002952.mm.1 | NM_001142647.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100002952.mm.1 | NM_001142647.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100002952.mm.1 | NR_024590.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100002952.mm.1 | NR_024590.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100002952.mm.1 | TC0100000380.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100002950.mm.1 | NM_001142647.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100002950.mm.1 | NM_001142647.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100002950.mm.1 | NR_024590.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100002950.mm.1 | NR_024590.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100002950.mm.1 | TC0100000380.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455553_PM_at | - | - |
| Mouse Expression Set 430 | 1455553_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455553_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455553_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220874 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220874 | - | - |

Tmem194b
Genomic Map