Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200038149.mm.1 |
NM_172673.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038149.mm.1 |
NM_172673.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038149.mm.1 |
TC0200004493.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200038145.mm.1 |
NM_172673.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038145.mm.1 |
NM_172673.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038145.mm.1 |
TC0200004493.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200038146.mm.1 |
NM_172673.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038146.mm.1 |
NM_172673.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038146.mm.1 |
TC0200004493.mm.1 |
MTA TC |