Assay Details
Gene Symbol: Slc25a24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172685.3 | NP_766273.1 | 4 - 5 | 567 | 127 | |
GenBank mRNA | AK032806.1 | - | 4 - 5 | 568 | 127 | |
AK146034.1 | - | 4 - 5 | 566 | 127 | ||
AK167778.1 | - | 4 - 5 | 558 | 127 | ||
AK168090.1 | - | 4 - 5 | 558 | 127 | ||
AK170598.1 | - | 4 - 5 | 559 | 127 | ||
AK187917.1 | - | 3 - 4 | 127 | |||
BC055369.1 | - | 4 - 5 | 556 | 30055307 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
