Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300009492.mm.1 |
NM_172685.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009492.mm.1 |
NM_172685.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009492.mm.1 |
TC0300001159.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009491.mm.1 |
NM_172685.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009491.mm.1 |
NM_172685.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009491.mm.1 |
TC0300001159.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009490.mm.1 |
NM_172685.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009490.mm.1 |
NM_172685.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009490.mm.1 |
TC0300001159.mm.1 |
MTA TC |